Learn More
Hoefer™ SUB6 Mini Submarine Combs
Accessories for SUB6 Mini Submarine
Brand: Hoefer™ SUB6-C1.5-8
Additional Details : Weight : 0.01000kg
Description
- Ranges from 8, 12 and 16 Wells
- 1, 1.5 and 2 mm Thickness
- 2.2, 2.5 and 4.5 mm Well Width
- Mini. 10, max. 40 μL Sample Volume
Specifications
SUB6 Mini Submarine | |
8 | |
0.15 cm | |
0.17 in. | |
0.19 in. |
Comb | |
0.05 in. | |
30 μL | |
0.45 cm | |
0.5 cm |