Learn More
Hoefer™ SUB6 Mini Submarine Combs
Accessories for SUB6 Mini Submarine
Brand:  Hoefer™ SUB6-C1-8
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12645806
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Ranges from 8, 12 and 16 Wells
  • 1, 1.5 and 2 mm Thickness
  • 2.2, 2.5 and 4.5 mm Well Width
  • Mini. 10, max. 40 μL Sample Volume
Specifications
Specifications
SUB6 Mini Submarine
8
0.1 cm
0.17 in.
0.19 in.
Comb
0.03 in.
20 μL
0.45 cm
0.5 cm