Learn More
Hoefer™ SUB6 Mini Submarine Combs
Accessories for SUB6 Mini Submarine
Brand: Hoefer™ SUB6-C2-12MC
Additional Details : Weight : 0.01000kg
Description
- Ranges from 8, 12 and 16 Wells
- 1, 1.5 and 2 mm Thickness
- 2.2, 2.5 and 4.5 mm Well Width
- Mini. 10, max. 40 μL Sample Volume
Specifications
SUB6 Mini Submarine | |
16 | |
0.2 cm | |
0.09 in. | |
0.19 in. |
Comb | |
0.07 in. | |
22 μL | |
0.25 cm | |
0.5 cm |