Learn More
Hoefer™ Tall Air-Cooled Vertical Electrophoresis Unit
Ideal for educational laboratories with multiple users
Brand:  Hoefer™ SE410-15-1.5
Additional Details : Weight : 0.80000kg
Product Code. 11935398
  • 1630.00€ / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • Compact, rugged and economical units
Specifications
Specifications
Air Cooled
60 mA
500 V
Tall Vertical Electrophoresis Unit
45°C