Learn More
Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles
Product Code. 11847221
Click to view available options
Lens Coating:
Hardcoat
Uncoated
Unit Size:
Each
Pack of 10
Product Code. 11847221
Brand: Sperian™ 1002221
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Offering high levels of comfort and support. Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles fit comfortably over prescription eyewear.
  • Practical and durable
  • Frosted side panels and browguard sections reduce glare/reflections
  • Curved temples and a raised nose bridge offer a precise fit
  • Available with or without hard lens coating
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001 / 2000, EN 166 1F
Lens Coating Hardcoat
Lens Tint Clear
Product Type Eye and Face Protection Spectacles
For Use With (Application) Over-the-glass spectacle, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling
Frame Color Transparent
No. per Pack 10/Pk.
Lens Material Polycarbonate