Learn More
Sperian™ Honeywell™ A2 Organic Vapour Filter Canisters for Respirators
Perfect shape to allow great vision in all situations. Sperian™ Honeywell™ A2 Organic Vapour Filter Canisters for Respirators can be used in places where the oxygen concentration is safely upper 17%.
Brand:  Sperian™ 1001577
Additional Details : Weight : 0.86400kg
Product Code. 11817171
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Can only be used with a half maks Valuair™, Premier™ or MX/PF or a full face Optifit Twin™ equipped with a T system
  • User checks installation via the audible click on its safety system
  • Coconut-based carbon charcoal impregnated for a high quality and safe level of filtration
  • Mechanical filtration membrane
  • Housing made of solid and resistant plastic material