Learn More
Honeywell™ Txl Pionner S3 Shoes
115.00€
Specifications
Certifications/Compliance | EN ISO 20345:2011, SRC Certified |
---|---|
For Use With (Application) | For professional use such as industry, green spaces, logistics |
Liner Material | Textile |
Material | Leather |
Sole Features | Full, ergonomical, removable and changeable insole, FLEXIUM™ composite puncture resistant mid-sole |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11817040
|
Honeywell™
6244411-40 |
40 |
115.00€
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
12665803
|
Honeywell™
6244411-39 |
39 |
115.00€
Each |
N/A | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
12685803
|
Honeywell™
6244411-47 |
47 |
115.00€
Each |
N/A | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
12675803
|
Honeywell™
6244411-46 |
46 |
115.00€
Each |
N/A | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Specifications
EN ISO 20345:2011, SRC Certified | |
Textile | |
Full, ergonomical, removable and changeable insole, FLEXIUM™ composite puncture resistant mid-sole | |
Freelium non-metallic | |
Yes |
For professional use such as industry, green spaces, logistics | |
Leather | |
Double density Polyurethane | |
Hydrocarbures, sole heat resistant up to 120°C, very good abrasion resistance | |
Shoes |