Learn More
Medicom™ Hopen™ Single Use Hygiene Gown
Medicom™ Hopen™ Single Use Hygiene Gown without pockets, is microporous and waterproof
135.40€
Specifications
| Certifications/Compliance | Manufactured under an ISO 9001 Certified Quality System |
|---|---|
| Certifications/Compliance (Europe) | EN 1149-5 |
| Color | White |
| Cuff Style | Elastic |
| Disposable | Yes |
| Product Code | Brand | Closure Type | Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Closure Type | Size | Price | Quantity & Availability | ||||
|
16524081
|
Medicom™
40.550/02 |
Press studs | Medium |
144.00€ 135.40€ / Pack of 50 Save 8.60€ 5% Off |
Estimated Shipment: 22-04-2026
Log in to see stock. |
||||
|
16534081
|
Medicom™
40.550/03 |
Press studs | Large |
144.00€ 135.40€ / Pack of 50 Save 8.60€ 5% Off |
Estimated Shipment: 22-04-2026
Log in to see stock. |
||||
|
16544081
|
Medicom™
40.550/04 |
Press studs | X-Large |
144.00€ 135.40€ / Pack of 50 Save 8.60€ 5% Off |
Estimated Shipment: 22-04-2026
Log in to see stock. |
||||
|
16554081
|
Medicom™
40.550/05 |
Press studs | 2X-Large |
144.00€ 135.40€ / Pack of 50 Save 8.60€ 5% Off |
Estimated Shipment: 22-04-2026
Log in to see stock. |
||||
Description
- Reduces contamination of the operator, but also sensitive products and processes in a protected atmosphere
- Protects against radioactive particles and infectious agents
- Microporous, Anti-Static Hygiene Gown is comfortable and light
- Polypropylene gown lined with laminated polyethylene for a waterproof, hydrophobic seal
- Available in mulitple sizes, and closure types
Specifications
| Manufactured under an ISO 9001 Certified Quality System | |
| White | |
| Yes | |
| Category 1 Personal Protective Equipment | |
| Polypropylene | |
| Individual, Bag | |
| Sewn |
| EN 1149-5 | |
| Elastic | |
| Microporous, Waterproof | |
| Unisex | |
| None | |
| Radioactive Particles, Infectious Agents | |
| Non-sterile |