Learn More
Macherey Nagel HPLC guard column EC 4/2NUCLEODUR 100-3 C18 ec lenght: 4 mm,
Brand:  Macherey Nagel 761931.20
HPLC guard column EC 4/2NUCLEODUR 100-3 C18 ec lenght: 4 mm,
Product Code. 12738972
  • 163.00€ / Pack of 1
Estimated Shipment: 25-09-2026