Learn More
Applied Biosystems™ HRM Software Module for QuantStudio™ 6 and 7 Systems
Product Code. 15471063
Click to view available options
Quantity:
10 licenses
Unit Size:
Each
Product Code. 15471063
Brand: Applied Biosystems™ 4486395
This software module expands the functionality of the QuantStudio™ 6 Flex and QuantStudio™ 7 Flex real-time PCR system software, allowing high resolution melting (HRM) analysis to be performed on PCR amplicons.
High Resolution Melt (HRM) analysis is a new, post-PCR analysis method used for identifying genetic variation in nucleic acid sequences. Simple and fast, this method is based on PCR melt (dissociation) curve techniques and is enabled by the recent availability of improved double-stranded DNA (dsDNA)-binding dyes along with next-generation real-time PCR instrumentation and analysis software. HRM analysis can discriminate DNA sequences based on their composition, length, GC content, or strand complementarity.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) QuantStudio™ 6 Flex, QuantStudio™ 7
Product Line QuantStudio
Software Type HRM Analysis Software
Product Type Software Module
For Use With (Application) Real Time PCR (qPCR)
Quantity 10 licenses