Learn More
HSP27, p Ser78 Antibody (2079A), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals MAB9355G
440.30 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18528689
  • 512.00€ / 100µL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
HSP27 Monoclonal antibody specifically detects HSP27 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HSP27
Monoclonal
DyLight 488
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Phosphopeptide containing the human HSP27 S78 site, Accession # P04792
100 μg
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience
3315
Store at 4°C in the dark.
IgG
Western Blot, Flow Cytometry, Immunocytochemistry
2079A