Learn More
Horst™ HSQ Model Flexible Heating Cable
Product Code. 10097011
Click to view available options
Length (Metric) Power Cord:
1m
3m
4m
5m
6m
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10097011
Brand: Horst™ 21001
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 1 Lm
Dimensions (L x W x H) 1 Lm
Electrical Requirements ∼230V Voltage
Model HSQ
Length (Metric) Power Cord 1m
Product Type Flexible Heating Cable