Learn More
Manutan Scouring green on roll, width: 10 cm, length: 300cm, colour: Green
Brand:  Manutan A018504
Encompass_Preferred
Scouring green on roll, width: 10 cm, length: 300cm, colour: Green
Product Code. 15583688
  • 34.97€ / Each
Only null left
Estimated Shipment: 18-12-2025