Learn More
Trajan Scientific and Medical™ HT Science Silicon Septa
Product Code. 15288613
Click to view available options
Diameter (Metric) Inner:
9 mm
11.5 mm
12.5 mm
Unit Size:
Pack of 24
Pack of 25
Product Code. 15288613
Brand: Trajan Scientific and Medical™ 041896
Only null left
Add to basket
Add to basket
Bleed and temperature optimized material.
  • Bleed and temperature optimized septa
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 400°C (330°C for 17mm size)
Specifications
Type HT
Diameter (Metric) Inner 9 mm
Quantity 25/Pk.