missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AATF Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0309

Additional Details : Weight : 0.01000kg

Product Code. 15276519

  • 130.00€ / 50µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

AATF Synthetic Peptide, PEP-0309, from Invitrogen.

This peptide corresponds to 12 amino acids near the carboxy terminus of human AATF.PEP-0309 can be used as a blocking peptide with polyclonal antibody PA5-20006.
Specifications

Specifications

26574
purified
Human AATF
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9NY61
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
AATF
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions