missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Arylsulfatase B/ARSB ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Human Arylsulfatase B/ARSB quantitates human Arylsulfatase B/ARSB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Arylsulfatase B/ARSB.
Arylsulfatase B encoded by this gene belongs to the sulfatase family. The arylsulfatase B homodimer hydrolyzes sulfate groups of N-Acetyl-D-galactosamine, chondriotin sulfate, and dermatan sulfate. The protein is targetted to the lysozyme. Mucopolysaccharidosis type VI is an autosomal recessive lysosomal storage disorder resulting from a deficiency of arylsulfatase B. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | 1110007C02Rik; AI480648; Arsb; arylsulfatase B; Arylsulfatase B (MPS VI); arylsulfatase B complex; arylsulfatase B regulation; arylsulfatase B structural; arylsulfatase B temporal regulation; As1; As-1; As-1 r; As1-r; As-1 s; As1-s; As-1 t; As1-t; ASB; Asr-1; Ast-1; G4S; MPS6; N-acetylgalactosamine-4-sulfatase |
| Assay Range | 0.32 to 75 ng/mL |
| Assay Sensitivity | 0.32 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 411 |
| Gene Symbol | ARSB |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction