missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ATAD3A Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP1273

Additional Details : Weight : 0.01000kg

Product Code. 15205279

  • 130.00€ / 50µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ATAD3A Synthetic Peptide, PEP-1273, from Invitrogen.

This peptide corresponds to 19 amino acids near the amino terminus of human ATAD3A.PEP-1273 can be used as a blocking peptide with polyclonal antibody PA5-21159.
Specifications

Specifications

55210
purified
Human ATAD3A
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9NVI7
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
ATAD3A
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions