Learn More
Gibco™ Human BDNF Recombinant Protein
Human BDNF Recombinant Protein
Brand: Gibco™ PHC7074
618.29 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Human BDNF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
- High purity—no interference from other proteins or contaminants
- High biological activity—more results with less protein
- Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media
High Purity Recombinant Bioactive Protein
Recombinant Human Brain-Derived Neurotrophic Factor is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High Biological Activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Brain-Derived Neurotrophic Factor (BDNF) specific activity is determined by measuring the dose-dependent induction of choline acetyl transferase activity in primary septal cell cultures.
Proven Compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
Specifications
Cell Culture | |
27 kDa | |
B-DNF | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
Neurotrophic Factors | |
Gibco™ |
627 | |
Sequential Chromatography | |
238 aa each (homodimer) | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Stem Cell Research, Differentiation | |
98 % |