missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TIM-1/KIM-1/HAVCR Fc Chimera Protein

Product Code. 18738328
Click to view available options
:
Quantity: 100μg
This item is not returnable. View return policy

Product Code. 18738328

Brand: R&D Systems™ 9319TM100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(268 aa)

The Recombinant Human TIM-1/KIM-1/HAVCR Fc Chimera Protein is derived from HEK293. The Recombinant Human TIM-1/KIM-1/HAVCR Fc Chimera Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAC39862
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 26762
Molecular Weight (g/mol) 55 kDa
Name TIM-1/KIM-1/HAVCR Fc Chimera Chimera Recombinant Protein
Quantity 100μg
Source Human embryonic kidney cell,HEK293-derived human TIM-1/KIM-1/HAVCR protein, (N-terminus) Human Tim-1/Kim-1/HAVCR (Ser21-Thr288) Accession &Num; AAC39862 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias CD365, HAVcr-1, HAVCR1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Common Name CD365; HAVCR1; HAVCR-1; HAVCRT cell immunoglobin domain and mucin domain protein 1; hepatitis A virus cellular receptor 1; Kidney injury molecule 1; KIM1; KIM-1; T-cell immunoglobulin and mucin domain-containing protein 1; TIM1; TIM-1; TIM-1TIM; TIM1TIMD-
Biological Activity 0.6 to 3.6μg/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.