missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Blimp-1 Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0433

Additional Details : Weight : 0.01000kg

Product Code. 15206649

  • 130.00€ / 50µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Blimp-1 Synthetic Peptide, PEP-0433, from Invitrogen.

Blimp-1 (B lymphocyte maturation protein-1) is a nuclear protein with five zinc fingers in its carboxy terminus. It is a transcriptional repressor that is both required and sufficient to trigger terminal differentiation of B lymphocytes and monocyte/macrophages. Blimp-1 is expressed in, and may be required for terminally differentiated epithelial cells, retinal neurons and other lineage cells. It has also been implicated in the repression of beta-interferon (IFN-beta) and c-myc gene expression. This peptide corresponds to 17 amino acids near the amino terminus of human Blimp-1.PEP-0433 can be used as a blocking peptide with polyclonal antibody PA5-20313.
Specifications

Specifications

639
purified
Human Blimp-1
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
O75626
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
PRDM1
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions