missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Breast Tissue Lysate (Adult Tumor)

Product Code. 18411172 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411172 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411172 Brand Novus Biologicals™ Brand No. NBP228008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adult Tumor Tissue Lysate

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method These lysates are proteomic discovery tools.Researchers should validate and optimize for individual use. Potential applications MAY include WB, immunoprecipitation, protein-protein interactions, ligand binding, ELISA. Note: For use in 1D and 2D gel electrophoresis, the addition of a denaturing gel loading buffer with reducing agents may be required.
Quantity 0.1 mg
Content And Storage Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles.
Target Breast
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.