missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BTBD12 (aa 717-812) Control Fragment Recombinant Protein

Product Code. 30196092
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196092

Brand: Invitrogen™ RP103627

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-65558 (PA5-65558, PA5-64270 (PA5-64270. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a structure-specific endonuclease subunit. The encoded protein contains a central BTB domain and it forms a multiprotein complex with the ERCC4(XPF)-ERCC1, MUS81-EME1, and SLX1 endonucleases, and also associates with MSH2/MSH3 mismatch repair complex, telomere binding complex TERF2(TRF2)-TERF2IP(RAP1), the protein kinase PLK1 and the uncharacterized protein C20orf94. The multiprotein complex is required for repair of specific types of DNA lesions and is critical for cellular responses to replication fork failure. The encoded protein acts as a docking platform for the assembly of multiple structure-specific endonucleases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IY92
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84464
Name Human BTBD12 (aa 717-812) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BTB (POZ) domain containing 12; BTB/POZ domain-containing protein 12; BTBD12; FANCP; KIAA1784; KIAA1987; MUS312; SLX4; SLX4 structure-specific endonuclease subunit; SLX4 structure-specific endonuclease subunit homolog; structure-specific endonuclease subunit SLX4
Common Name BTBD12
Gene Symbol SLX4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LIQYVNNEGFSAVEDGVLTQRVLLGDVSTEAARTFLHYLYTADTGLPPGLSSELSSLAHRFGVSELVHLCEQVPIATDSEGKPWEEKEAENCESRA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.