missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C1orf19 Full-length ORF (NP_443197.1, 1 a.a. - 171 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00116461-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157343

  • 528.00€ / 25µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

tRNA splicing is a fundamental process required for cell growth and division. SEN15 is a subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns, the first step in tRNA splicing (Paushkin et al., 2004 [PubMed 15109492]).[supplied by OMIM]

Sequence: MEERGDSEPTPGCSGLGPGGVRGFGDGGGAPSWAPEDAWMGTHPKYLEMMELDIGDATQVYVAFLVYLDLMESKSWHEVNCVGLPELQLICLVGTEIEGEGLQTVVPTPITASLSHNRIREILKASRKLQGDPDLPMSFTLAIVESDSTIVYYKLTDGFMLPDPQNISLRR
Specifications

Specifications

NP_443197.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1orf19/sen15
TSEN15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
116461
C1orf19 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEERGDSEPTPGCSGLGPGGVRGFGDGGGAPSWAPEDAWMGTHPKYLEMMELDIGDATQVYVAFLVYLDLMESKSWHEVNCVGLPELQLICLVGTEIEGEGLQTVVPTPITASLSHNRIREILKASRKLQGDPDLPMSFTLAIVESDSTIVYYKLTDGFMLPDPQNISLRR
RUO
TSEN15
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions