missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCP-3/CCL7 Human Uncoated ELISA Kit with Plates

Product Code. 15500977
Change view
Click to view available options
Quantity:
2 x 96 Tests
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15500977 2 x 96 Tests Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15500977 Supplier Invitrogen™ Supplier No. 885070022

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Includes

  • Capture Antibody Concentrate
  • Detection Antibody Concentrate
  • Human CCL7( MCP-3) Standard (lyophilized): 100pg/mL upon reconstitution
  • Coating Buffer Concentrate
  • Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
  • Sample Diluent Z
  • pre-titrated Streptavidin-HRP
  • Tetramethylbenzidine (TMB) Substrate Solution

Uncoated ELISA Kit with Plates

The Human Monocyte Chemoattractant Protein 3 (CCL7) MCP-3) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human MCP-3. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

This is a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80098
Assay Range 0.3125-20 pg/mL
Assay Sensitivity 0.3125 pg/mL
Conjugate Biotin
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6354
Gene Symbol FIC,MARC,MCP-3,MCP3,NC28,SCYA6,SCYA7
Kit Contents Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
Label or Dye HRP
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Storage Requirements 2°C to 8°C
Synonym FIC, MARC, Monocyte chemotactic protein 3
Target Human
Test Time 1 hr. 45 min.
Total Assay Time 24 hr. 30 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.