missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD163 DuoSet ELISA

Product Code. 30001756
Click to view available options
Quantity:
5 Plates
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 30001756

Brand: R&D Systems™ DY160705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Human CD163 Capture Antibody; Human CD163 Detection Antibody; Human CD163 Standard; Streptavidin-HRP

Human CD163 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CD163 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CD163.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 Plates
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.