missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CENTG2 Full-length ORF (AAH60814.1, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00116987-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157383

  • 349.00€ / 10µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CENTG2 belongs to an ADP-ribosylation factor GTPase-activating (ARF-GAP) protein family involved in membrane traffic and actin cytoskeleton dynamics (Nie et al., 2002 [PubMed 12388557]).[supplied by OMIM]

Sequence: MNYQQQLANSAAIRAEIQRFESVHPNIYSIYELLERVEEPVLQNQIREHVIAIEDAFVNSQEWTLSRSVPELKVGIVGNLASGKSALVHRYLTGTYVQEESPEGGRFKKEIVVDGQSYLLLIRDEGGPPEAQFAMWVDAVIFVFSLEGEISFQTVYHYYSRMANYRNTSEIPLVLVGTQDAISSANPRVIDDARARKLSNDLKRCTYYETCATYGLNVERVFQDVAQKIVATRKKQQLSIGPCKSLPNSPSHSSVCSAQVSAVHISQTSNGGGSLSDYSSSVPSTPSTSQKELRIDVPPTANTPTPVRKQSKRRSNLFTSRKGSDPDKEKKGLESRADSIGSGRAIPIKQGLPFFVLALTASTYLRPAGARARQSSPWPGRAEGWSDQPALC
Specifications

Specifications

AAH60814.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
69.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
AGAP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
116987
CENTG2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CENTG2/GGAP1/KIAA1099/MGC71657
AGAP1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions