missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human COL5A3 Control Fragment Recombinant Protein

Product Code. 30182524
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182524

Brand: Invitrogen™ RP98883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (52%), Rat (52%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-144797 (PA5-144797, PA5-83902 (PA5-83902. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P25940
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 50509
Name Human COL5A3 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias COL5A3; Collagen 5; Collagen alpha-3(V) chain; collagen type V alpha 3; collagen, type V, alpha 3; collagen, type V, alpha 4; pro-(alpha)3(V) collagen; Pro-alpha3(V); procollagen, type V, alpha 3
Common Name COL5A3
Gene Symbol COL5A3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence STDIPKTETPAPNLPPTPTPLVVTSTVTTGLNATILEGSLDPDSGTELGTLETKAAREDEEGDDSTMGPDFRAAEYPSRTQFQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.