missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CRIM1 ELISA Kit

Product Code. 17184403
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17184403

Brand: Invitrogen™ EH129RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human CRIM1 quantitates human CRIM1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRIM1.

CRIM1 (cysteine-rich motor neuron 1), a glycosylated type I transmembrane protein, plays a role in tissue development i.e. capillary formation and maintenance during angiogenesis. It contains an N-terminal IGF-binding protein-like motif and six von Willebrand-like cysteine-rich repeats (CRRs) in its extracellular domain. CRIM1 interacts with BMP4 and BMP7 via the CRRs and functions as an antagonist. CRIM1 is developmentally expressed in a number of tissues including the pancreas, kidney, placenta, brain and blood vessels. CRIM1 may participate in CNS and placental development by interacting with growth factors involved in motor neuron differentiation and survival.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AU015004; Crim1; CRIM-1; cysteine rich transmembrane BMP regulator 1; cysteine rich transmembrane BMP regulator 1 (chordin like); cysteine rich transmembrane BMP regulator 1 (chordin-like); cysteine-rich motor neuron 1; cysteine-rich motor neuron 1 protein; Cysteine-rich repeat-containing protein S52; Processed cysteine-rich motor neuron 1 protein; S52; UNQ1886/PRO4330
Assay Range 0.041 to 10 ng/mL
Assay Sensitivity 0.041 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 51232
Gene Symbol CRIM1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CRIM1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.