Learn More
R&D Systems™ Human Cystatin C Magnetic Luminex™ Performance Assay
Product Code. 18471800
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18471800
Brand: R&D Systems™ LHK1196
For use with the magnetic Kidney Biomarker Panel
  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY
Specifications
Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target Cystatin C
Bead Type Magnetic
Assay Sensitivity 57.1pg/mL
Assay Range 23.00 - 16500pg/mL
Gene ID (Entrez) 1471
Gene Symbol CST3
Sample Type Serum, Plasma (EDTA, Heparin), Urine
Sample Volume Serum (10μL),EDTA Plasma (10μL),Heparin Plasma (10μL),Urine (10μL)
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.