Learn More
Invitrogen™ Human Cytoglobin (aa 44-173) Control Fragment Recombinant Protein
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53637 (PA5-53637. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spezifikation
Spezifikation
| Accession Number | Q8WWM9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 114757 |
| Name | Human Cytoglobin (aa 44-173) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 3110001K20Rik; Cygb; Cytoglobin; HGb; Histoglobin; Staap; STAP; stellate cell activation associated protein; stellate cell activation-associated protein |
| Common Name | Cytoglobin |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.