missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DLEC/CLEC4C/BDCA-2 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human DLEC/CLEC4C/BDCA-2 quantitates human DLEC/CLEC4C/BDCA-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DLEC/CLEC4C/BDCA-2.
This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
Specifications
| Accession Number | BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150 |
| Assay Range | 0.082 to 20 ng/mL |
| Assay Sensitivity | 0.082 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 170482 |
| Gene Symbol | CLEC4C |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction