missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DTX1 Partial ORF (NP_004407, 381 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001840-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157364

  • 349.00€ / 10µg
Estimated Shipment: 10-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Studies in Drosophila have identified this gene as encoding a positive regulator of the Notch-signaling pathway. The human gene encodes a protein of unknown function; however, it may play a role in basic helix-loop-helix transcription factor activity. [provided by RefSeq]

Sequence: TKKKHLKKSKNPEDVVRRYMQKVKNPPDEDCTICMERLVTASGYEGVLRHKGVRPELVGRLGRCGHMYHLLCLVAMYSNGNKDGSLQCPT
Specifications

Specifications

NP_004407
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TKKKHLKKSKNPEDVVRRYMQKVKNPPDEDCTICMERLVTASGYEGVLRHKGVRPELVGRLGRCGHMYHLLCLVAMYSNGNKDGSLQCPT
RUO
DTX1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1840
DTX1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
hDx-1
DTX1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions