missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Endocan/ESM-1 ELISA Kit (Chemiluminescence)

Product Code. 18683360 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18683360

Brand: Novus Biologicals™ NBP268154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Chemiluminescence)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.81 to 500 pg/mL
Assay Sensitivity 4.69 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Human ESM1 in samples. No significant cross-reactivity or interference between Human ESM1 and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 11082
Gene Symbol ESM1
Quantity 1 Kit
Research Discipline Angiogenesis, Cancer
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym endocan, endothelial cell-specific molecule 1, ESM-1
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.