missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ENOX2 (aa 441-563) Control Fragment Recombinant Protein

Product Code. 30199198
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
30199198 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 30199198

Brand: Invitrogen™ RP91909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51398 (PA5-51398. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a tumor-specific member of the ECTO-NOX family of genes that encode cell surface NADH oxidases. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The protein also displays prion-like properties. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16206
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10495
Name Human ENOX2 (aa 441-563) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias APK1; APK1 antigen; Cova1; cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; enox2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; RGD1563643; tNOX; tumor-associated hydroquinone oxidase; zmp:0000001197
Common Name ENOX2
Gene Symbol Enox2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KVHREDDPNKEQQLKLLQQALQGMQQHLLKVQEEYKKKEAELEKLKDDKLQVEKMLENLKEKESCASRLCASNQDSEYPLEKTMNSSPIKSEREALLVGIISTFLHVHPFGASIEYICSYLHR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.