missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EPS8 Full-length ORF (NP_004438.3, 1 a.a. - 822 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002059-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157497

  • 349.00€ / 10µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq]

Sequence: MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH
Specifications

Specifications

NP_004438.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPS8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
2059
EPS8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH
RUO
EPS8
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions