missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Erythropoietin/EPO Quantikine ELISA Kit, RUO

Product Code. 30000729
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 30000729

Brand: R&D Systems™ DEPRU0

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Human Erythropoietin Quantikine ELISA Kit, RUO from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility

The Human Erythropoietin Quantikine ELISA Kit, RUO quantifies human Erythropoietin in serum (100 μL), edta plasma (100 μL).
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA
Specificity Natural and Recombinant Human Epo
Gene ID (Entrez) 2056
Host Species Human
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Target Erythropoietin/EPO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.