missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FABP6 ELISA Kit

Product Code. 17124463
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17124463

Brand: Invitrogen™ EH179RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human FABP6 quantitates human FABP6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FABP6.

This gene encodes the ileal fatty acid binding protein. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP6 and FABP1 (the liver fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. Transcript variants generated by alternate transcription promoters and/or alternate splicing have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number FABP6; fatty acid binding protein 6; fatty acid binding protein 6, ileal; fatty acid binding protein 6, ileal (gastrotropin); fatty acid-binding protein 6; Gastrotropin; GT; I-15 P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ileal bile acid binding protein; ileal bile acid-binding protein; ileal lipid-binding protein; ILLBP; illeal lipid-binding protein; intestinal 15 kDa protein; Intestinal bile acid-binding protein
Assay Range 1.64 to 400 ng/mL
Assay Sensitivity 1.64 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2172
Gene Symbol FABP6
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target FABP6
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.