missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-19, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantity:
250 μg
5 μg
Unit Size:
250µg
5µg
Descripción
Recombinant Human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF19, Fibroblast growth factor 19, is a protein that in humans is encoded by the FGF19 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. The FGF19 gene is mapped to 11q13.3. The deduced 216-amino acid FGF19 protein contains asignal sequence and 2 cysteine residues that are conserved in the FGF family. Expression of this gene was detected only in fetal but not adult brain tissue. Synergistic interaction of the chick homolog and Wnt-8c has been shown to be required for initiation of inner ear development. FGF19 stimulates hepatic protein and glycogen synthesis but does not induce lipogenesis. The effects of FGF19 are independent of the activity of either insulin or the protein kinase Akt and, instead, are mediated through a mitogen-activated protein kinase signaling pathway that activates components of the protein translation machinery and stimulates glycogen synthase activity. The orthologous protein in mouse is FGF15, which shares about 50% amino acid identity and has similar functions. Together they are often referred to as FGF15/19.
Especificaciones
Especificaciones
| Accession Number | O95750 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 9965 |
| Molecular Weight (g/mol) | 21.8 kDa |
| Name | Human FGF-19, Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 5 μg |
| Regulatory Status | RUO |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido