missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Frizzled-2 (aa 136-168) Control Fragment Recombinant Protein

Product Code. 30180944
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180944

Brand: Invitrogen™ RP99941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the wingless type MMTV integration site family of signaling proteins. This gene encodes a protein that is coupled to the beta-catenin canonical signaling pathway. Competition between the wingless-type MMTV integration site family, member 3A and wingless-type MMTV integration site family, member 5A gene products for binding of this protein is thought to regulate the beta-catenin-dependent and -independent pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14332
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2535
Name Human Frizzled-2 (aa 136-168) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AL033370; AW456835; Dfrizzled2; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 2; frizzled 2, seven transmembrane spanning receptor; frizzled class receptor 2; frizzled family receptor 2; frizzled homolog 2; frizzled homolog 2 (Drosophila); frizzled-10; Frizzled-2; Fz10; fz-10; Fz2; Fz-2; Fzd10; Fzd2; fzE2; hFz2; Mfz10; Mfz10a; mFz2; rFz2
Common Name Frizzled-2
Gene Symbol Fzd2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LRCEHFPRHGAEQICVGQNHSEDGAPALLTTAP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.