missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Galectin-9 ELISA Kit

Product Code. 17184959
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17184959

Brand: Invitrogen™ EH206RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Galectin-9 quantitates human Galectin-9 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Galectin-9.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; Ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; Tumor antigen HOM-HD-21; UAT; Urate transporter/channel; urate transporter/channel protein
Assay Range 36.86 to 9,000 pg/mL
Assay Sensitivity 36 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3965
Gene Symbol Lgals9
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Galectin 9
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.