missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GASP-1 ELISA Kit

Product Code. p-7219395
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17114969

Brand: Invitrogen™ EH209RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human GASP-1 quantitates human GASP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GASP-1.

Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2210415K24Rik; 3110031O14Rik; C87852; G protein-coupled receptor associated sorting protein 1; GASP; GASP1; GASP-1; GPRASP1; G-protein coupled receptor-associated sorting protein 1; Kiaa0443; Per1 interacting protein; per1-interacting protein; Pips
Assay Range 20 to 5,000 pg/mL
Assay Sensitivity 20 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 9737
Gene Symbol GPRASP1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GPRASP1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.