missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPR56 ELISA Kit

Product Code. 17174513
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17174513

Brand: Invitrogen™ EH228RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human GPR56 quantitates human GPR56 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GPR56.

GPR56 is a member of the G protein-coupled receptor family. The protein contains 7 transmembrane domains and a mucin-like domain in the N-terminal region. The gene is implicated in the regulation of brain cortical patterning. The protein binds specifically to transglutaminase 2 in the extracellular space. Expression of this gene is downregulated in melanoma cell lines, and overexpression of this gene can suppress tumor growth and metastasis. Mutations in this gene result in bilateral frontoparietal polymicrogyria. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 7-transmembrane protein with no EGF-like N-terminal domains-1; Adgrg1; ADGRG1 CT; ADGRG1 C-terminal fragment; ADGRG1 NT; ADGRG1 N-terminal fragment; ADGRG1-CT; adhesion G protein-coupled receptor G1; adhesion G-protein coupled receptor G1; BFPP; BPPR; Cyt28; DKFZp781L1398; G protein-coupled receptor 56; Gpr56; GPR56 7 TM; GPR56 CT; GPR56 C-terminal fragment; GPR56 extracellular subunit; GPR56 NT; GPR56 N-terminal fragment; GPR56 seven-transmembrane subunit; GPR56 subunit alpha; GPR56 subunit beta; G-protein coupled receptor 56; Protein TM7XN1; secretin receptor; serpentine receptor cyt28; testicular tissue protein Li 77; TM7LN4; TM7XN1; UNQ540/PRO1083
Assay Range 16 to 4,000 pg/mL
Assay Sensitivity 16 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 9289
Gene Symbol ADGRG1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GPR56
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.