missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GPR56 ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EH228RBX10
Additional Details : Weight : 2.50000kg
Description
GPR56 is a member of the G protein-coupled receptor family. The protein contains 7 transmembrane domains and a mucin-like domain in the N-terminal region. The gene is implicated in the regulation of brain cortical patterning. The protein binds specifically to transglutaminase 2 in the extracellular space. Expression of this gene is downregulated in melanoma cell lines, and overexpression of this gene can suppress tumor growth and metastasis. Mutations in this gene result in bilateral frontoparietal polymicrogyria. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Q9Y653 | |
16 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
ADGRG1 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
16 to 4,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
9289 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL|Plasma, 50 μL|Serum, 50 μL | |
GPR56 | |
4 hr 45 min |