missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Grik2 Synthetic Peptide
Description
Grik2 Synthetic Peptide, PEP-0566, from Invitrogen.
Glutamate receptors are the primary excitatory neurotransmitter receptors in the mammalian brain and are activated during various normal neurophysiological processes. The kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels, includes this gene product. The subunit encoded by this gene is edited at multiple sites within the first and second transmembrane domains, which is thought to alter the structure and function of the receptor complex. Furthermore, different isoforms of this gene have been described due to alternatively spliced transcript variants. Mutations in this gene have been linked to autosomal recessive cognitive disability. GRIK2-associated diseases include Neurodevelopmental Disorder with Impaired Language and Ataxia and with or without Seizures and Intellectual Developmental Disorder, Autosomal Recessive 6.
Specifications
Specifications
| Gene ID (Entrez) | 2898 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q13002 |
| Common Name | Human Grik2 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?