missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRIK5 Synthetic Peptide

Product Code. 15388710
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
15388710 50 μg 50µg
1 options
This item is not returnable. View return policy

Product Code. 15388710

Brand: Invitrogen™ PEP0568

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Grik5 Synthetic Peptide, PEP-0568, from Invitrogen.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. Grik5, also known as kainate-preferring glutamate recptor subunit KA2, belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. Grik5 is highly homologous to the related ionotrophic glutamate receptor Grik4 (also known as KA1). Like Grik4, Grik5 does not form homomeric channels, but instead forms heteromers with Grik2. In Grik2- but not Grik1-null mice, Grik5 surface expression is greatly reduced in neurons, indicating that Grik2/Grik5 heteromers are required for exit from the endoplasmic reticulum to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2901
Species Human
Purification Method purified
Accession Number Q16478
Common Name Human GRIK5
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol GRIK5
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.