missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HBXAP Partial ORF (NP_057662.3, 1343 a.a. - 1441 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051773-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157316

  • 349.00€ / 10µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HBXAP is involved in transcription repression, transcription coactivation when associated with hepatitis B virus X protein (HBX), and chromatin remodeling and spacing when associated with SNF2H (MIM 603375).[supplied by OMIM]

Sequence: IESDEEEDFENVGKVGSPLDYSLVDLPSTNGQSPGKAIENLIGKPTEKSQTPKDNSTASASLASNGTSGGQEAGAPEEEEDELLRVTDLVDYVCNSEQL
Specifications

Specifications

NP_057662.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IESDEEEDFENVGKVGSPLDYSLVDLPSTNGQSPGKAIENLIGKPTEKSQTPKDNSTASASLASNGTSGGQEAGAPEEEEDELLRVTDLVDYVCNSEQL
RUO
RSF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51773
HBXAP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HBXAP/RSF-1/XAP8/p325
RSF1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions