missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HGF Quantikine ELISA Kit

Product Code. p-7145526
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
Unit Size:
1 set
Pack of 96
This item is not returnable. View return policy

Product Code. 18742254

Brand: R&D Systems™ DHG00B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human HGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human HGF Quantikine ELISA Kit quantifies human HGF in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul).

1 Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 156.00 - 10000 pg/mL
Assay Sensitivity 24.1 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human HGF
For Use With (Application) ELISA
Gene ID (Entrez) 3082
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL),Citrate Plasma (50 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Target HGF
Total Assay Time 4 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.