missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IFN-beta DuoSet ELISA
Click to view available options
Format:
96-well Plates
Unit Size:
Pack of 5
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human IFN-beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IFN-beta.
Specifications
Specifications
| Product Type | ELISA Kit |
| Species | Human |
| Binding Target | IFN-beta |
| Format | 96-well Plates |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3456 |
| Sample Type | Cell Culture Supernatant, Serum, Plasma |
| Synonym | Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction