Learn More
STEMCELL Technologies Human IL-1 beta ELISA Kit
Product Code. p-7217513
Encompass_Preferred
Click to view available options
Quantity:
2 Plates
Unit Size:
Pack of 2
Product Code. 17128011
Brand: STEMCELL Technologies 02004
For detection and measurement of human interleukin 1 beta
  • The Human Interleukin 1 beta (IL-1β) ELISA Kit is designed for the quantitative detection and measurement of human IL-1β in biological fluids such as serum, plasma, and cell culture supernatants
  • IL-1β is secreted by monocytes, macrophages, and dendritic cells
  • IL-1β is a proinflammatory cytokine that recruits immunocompetent cells to affected sites by stimulating the expression of adhesion molecules
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Specifications
Description Dendritic Cells; Macrophages; Monocytes
Quantity 2 Plates