missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-1 R3/IL-1 R Acp ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human IL-1 R3/IL-1 R Acp quantitates human IL-1 R3/IL-1 R Acp in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-1 R3/IL-1 R Acp.
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
Specifications
Specifications
| Accession Number | 6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1 R 3; IL 1R3; IL 1 RAcP; IL 1 RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1 R-3; IL1RAcP; IL-1 RAcP; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain |
| Assay Range | 18 to 4,000 pg/mL |
| Assay Sensitivity | 18 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 3556 |
| Gene Symbol | IL1RAP |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction